Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P4167

Sigma-Aldrich

Potassium sulfate

ACS reagent, ≥99.0%, powder

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
K2SO4
CAS Number:
Molecular Weight:
174.26
EC Number:
MDL number:
UNSPSC Code:
12352300

grade

ACS reagent

Assay

≥99.0%

form

powder

impurities

≤0.01% Insoluble matter
≤5 ppm Nitrogen compounds (as N)

pH

5.5-8.5 (25 °C, 5% in solution)

anion traces

chloride (Cl-): ≤0.001%

cation traces

Ca: ≤0.01%
Fe: ≤5 ppm
Mg: ≤0.005%
Na: ≤0.02%
heavy metals (as Pb): ≤5 ppm

SMILES string

[K+].[K+].[O-]S([O-])(=O)=O

InChI

1S/2K.H2O4S/c;;1-5(2,3)4/h;;(H2,1,2,3,4)/q2*+1;/p-2

InChI key

OTYBMLCTZGSZBG-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service