Skip to Content
MilliporeSigma
All Photos(1)

Documents

1.12553

Sigma-Aldrich

Coomassie Brilliant blue R 250 (C.I. 42660)

for electrophoresis Trademark of Imperial Chemical Industries PLC

Synonym(s):

Coomassie Brilliant blue R 250 (C.I. 42660), protein stain

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C45H44N3NaO7S2
CAS Number:
Molecular Weight:
825.97
MDL number:
UNSPSC Code:
41105305
EC Index Number:
228-060-5
NACRES:
NC.01

Quality Level

potency

>5000 mg/kg LD50, oral (Rat)

technique(s)

protein staining: suitable

pH

6.2 (25 °C, 10 g/L in H2O)

mp

174-180 °C

solubility

20 g/L

bulk density

250 kg/m3

εmax

≥300 at 554-563 nm at 0.025 g/L (pH 7.0)

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

2-30°C

InChI

1S/C45H45N3O7S2.Na/c1-4-47(31-33-9-7-11-43(29-33)56(49,50)51)40-23-15-36(16-24-40)45(35-13-19-38(20-14-35)46-39-21-27-42(28-22-39)55-6-3)37-17-25-41(26-18-37)48(5-2)32-34-10-8-12-44(30-34)57(52,53)54;/h7-30,46H,4-6,31-32H2,1-3H3,(H-,49,50,51,52,53,54);/q;+1/p-1

InChI key

HAEGGLCXLFWTBE-UHFFFAOYSA-M

Analysis Note

Identity (UV/VIS-Spectrum): passes test
Absorption maximum λmax. (buffer pH 7.0): 554 - 563 nm
Spec. Absorptivity A 1%/1cm (λmax; 0.025 g/l; buffer pH 7.0; calc. on dried substance): ≥ 300
TLC-Test: passes test
Loss on drying (110 °C): ≤ 5 %
Suitability for electrophoresis: passes test

Legal Information

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service