Documents
MTCC3220SGL
MTC Bio ClearSeal Graduated Screw Cap Microtubes
capacity 2.0 mL, sterile, cap, natural(looped), self standing, pack of 1000 ea (20 x bags 50 ea)
Synonym(s):
centrifuge tube, screw-cap tube
About This Item
UNSPSC Code:
41121703
NACRES:
NB.15
material
(self standing star foot)
cap (screw top loop )
clear
polypropylene
sterility
sterile
feature
cap natural(looped)
packaging
pack of 1000 ea (20 x bags 50 ea)
manufacturer/tradename
MTC Bio C3220-SGL
parameter
22,000 × g speed
capacity
2.0 mL
diam. × L
13 mm × 49.2 mm
General description
ClearSeal screw-cap microtubes, 2.0mL, w/ O-ring & loop-cap, sterile, printed graduations, self-standing, 20 resealable bags of 50 tubes, 1000/cs
Screw-cap microtubes include assembled caps with silicone O-rings that assure a leakproof seal from -86°C to +121°C and can be centrifuged at up to 22,000×g.
Features and Benefits
  • Ultra-clear polypropylene
  • Packaged in resealable bags of 50
  • Leak-proof O-ring seal
  • Sterile, RNase, DNase & Pyrogen free
  • White, printed graduations
Legal Information
ClearSeal is a trademark of MTC Bio
MTC is a trademark of MTC Bio