MTCC2051
MTC™ Bio Locktop Style Microtube
capacity 1.5 mL, pack of 500 ea
Synonym(s):
centrifuge tube, plastic tube
All Photos(1)
About This Item
Recommended Products
material
clear
polypropylene
round
sterility
non-sterile
feature
graduation
packaging
pack of 500 ea
manufacturer/tradename
MTC™ Bio C2051
capacity
1.5 mL
suitability
suitable for PCR
Related Categories
General description
MTC™ Bio 0.5 mL microtube come with Locktop style caps. Designed for secure sample storage, the locktop caps ensure a tight seal to prevent any leakage or contamination. These microtubes are ideal for various laboratory applications such as sample storage, PCR, and centrifugation. Their robust construction enables them to withstand high speeds in centrifugation processes without compromising sample integrity. The clear polypropylene material allows for easy visual inspection of the samples. These microtubes are the perfect choice for any lab seeking reliable and convenient sample containment solutions.
Microtube w/ cap, 1.5ml, Locktop style cap, 500/pk
Features and Benefits
- Compatible with standard equipment
- Centrifuge at up to 28,000xg
- RNase, DNase & pyrogen free
- Unsurpassed Clarity
Legal Information
MTC is a trademark of MTC Bio