Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MTCC2000R

MTC Bio SureSeal S Microcentrifuge Tube

capacity 1.5 mL, sterile, red, pack of 500 ea

Synonym(s):

centrifuge tube, plastic tube

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121703
NACRES:
NB.15

material

conical bottom
polypropylene
red

sterility

sterile

packaging

pack of 500 ea

manufacturer/tradename

MTC Bio C2000-R

parameter

28,000 × g speed

capacity

1.5 mL

Looking for similar products? Visit Product Comparison Guide

General description

Designed for a broad range of research applications, including enzyme digests, DNA isolation/purification, centrifugation, incubation, and general sample/reagent storage. SureSeal(TM) S tubes are manufactured under clean room conditions, using only BioClear co-polymer, an advanced polypropylene that maximizes clarity, centrifugal strength (tested up to 28,000×g) and the usable temperature range (-90°C to +121°C). The full-sized tube cap (with frosted marking surface), combined with the unique ring and groove closure, enhances the security of the liquid-tight seal, while minimizing the required locking forces for a more ergonomic opening/closing of the tube caps.
Microtube w/ cap, 1.5ml, red, sterile, w/ self-standing bag & Stop-Pops, 500/pk

Features and Benefits

  • Sterile comes standard – by electron beam radiation
  • Self-Standing bag – with gusseted bottom
  • Stop-Pops supplied – 10 cap locks in every bag

Legal Information

MTC is a trademark of MTC Bio
SecureSeal is a trademark of Grace Bio-Labs, Inc.
Stop-Pops is a trademark of MTC Bio
SureSeal S is a trademark of MTC Bio

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service