Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MSST0013

Sigma-Aldrich

SILuProt AMBP Alpha-1 Microglycoprotein human

recombinant, expressed in HEK 293 cells, SIL MS Protein Standard, 13C and 15N-labeled

Synonym(s):

Alpha-1-microglobulin, Complex-forming glycoprotein heterogeneous in charge

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23201100
NACRES:
NA.12
Pricing and availability is not currently available.

biological source

human

Quality Level

recombinant

expressed in HEK 293 cells

tag

His tagged

Assay

≥98% (SDS-PAGE)

form

lyophilized powder

potency

≥98% Heavy amino acids incorporation efficiency by MS

technique(s)

mass spectrometry (MS): suitable

suitability

suitable for mass spectrometry (standard)

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... AMBP(259)

Related Categories

General description

SILu Prot AMBP is a recombinant, stable isotope-labeled human AMBP which incorporates [13C6, 15N4]-Arginine and [13C6, 15N2]-Lysine. Expressed in human 293 cells, it is designed to be used as an internal standard for bioanalysis of AMBP in mass-spectrometry. SILu Prot AMBP is a monomer of 204 amino acids (including C-terminal polyhistidine and flag tags), with a molecular weight of ~23 kDa.

Biochem/physiol Actions

AMBP is synthesized by the liver, with approximately half of the circulating protein complexed to IgA. The free form is readily filtered by the glomerulus and reabsorbed by proximal tubule cells. AMBP has been found to be a sensitive biomarker for proximal tubular dysfunction even in the early phase of injury when no histologic damage is observable. In addition, urinary AMBP has been proposed to be a useful marker of tubular dysfunction even in low-gestational-age preterm infants, a population at high risk for AKI (Acute Kidney Injury).

Sequence

GPVPTPPDNIQVQENFNISRIYGKWYNLAIGSTCPWLKKIMDRMTVSTLVLGEGATEAEISMTSTRWRKGVCEETSGAYEKTDTDGKFLYHKSKWNITMESYVVHTNYDEYAIFLTKKFSRHHGPTITAKLYGRAPQLRETLLQDFRVVAQGVGIPEDSIFTMADRGECVPGEQEPEPILIPRVDYKDDDDKGHHHHHHHHGGQ

Physical form

Supplied as a lyophilized powder containing phosphate buffered saline.

Legal Information

SILu is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service