Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

860853P

Avanti

16:0 Azido Coenzyme A

Avanti Research - A Croda Brand, powder

Synonym(s):

16-azidohexadecanoyl Coenzyme A (ammonium salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C37H74N13O17P3S
CAS Number:
Molecular Weight:
1098.05
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (860853-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

click reagents

shipped in

dry ice

storage temp.

−20°C

SMILES string

O[C@@](C(NCCC(NCCSC(CCCCCCCCCCCCCCCN=[N+]=[N-])=O)=O)=O)(C(C)(COP([O-])(OP([O-])(OC[C@H]([C@H]1OP([O-])(O)=O)O[C@H]([C@@H]1O)N2C3=C(C(N)=NC=N3)N=C2)=O)=O)C)[H].[NH4+].[NH4+].[NH4+]

InChI

1S/C37H65N10O17P3S.3H3N/c1-37(2,32(51)35(52)41-19-17-27(48)40-20-21-68-28(49)16-14-12-10-8-6-4-3-5-7-9-11-13-15-18-45-46-39)23-61-67(58,59)64-66(56,57)60-22-26-31(63-65(53,54)55)30(50)36(62-26)47-25-44-29-33(38)42-24-43-34(29)47;;;/h24-26,30-32,36,50-51H,

InChI key

IRKUQXJWDGYOIA-LFYAMDRWSA-N

General description

Azido Coenzyme A is a derivative of coenzyme A.

Packaging

pkg of 1 x 1mg; 5mL glass amber screw cap vial (860853P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

One-step transformation of coenzyme A into analogues by transamidation
Sanichar R and Vederas J C
Organic Letters, 19(8), 1950-1953 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service