Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MAK518

Sigma-Aldrich

ADP Assay Kit

sufficient for 100 fluorometric tests

Synonym(s):

ADP Detection Kit

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
$654.00

$654.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 KIT
$654.00

About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

$654.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

detection method

fluorometric

relevant disease(s)

hematological disorder

shipped in

wet ice

storage temp.

−20°C

General description

Adenosine diphosphate (ADP) is the product of ATP dephosphorylation by ATPases. ADP can be converted back to ATP by ATP synthases. ADP levels regulate several enzymes involved in intermediary metabolism. Conventionally, ADP levels are measured by luciferase/luciferin mediated assays after ADP is converted to ATP. However, since these assays require measurement of ATP in the sample before conversion of ADP to ATP, if the nascent ATP concentration is significantly higher than the ADP concentration, the ATP signal will drown out the ADP signal.

Features and Benefits

  • As low as 0.1 µM ADP can be quantified.
  • Compatible with high-throughput handling systems.

Suitability

The kit is suitable for ADP determination in cells and other biological samples.

Principle

The ADP Assay Kit provides a convenient fluorometric means to measure ADP level even in the presence of ATP. In the assay, ADP is converted to ATP and pyruvate. The generated pyruvate is then quantified by a fluorometric method at λEx = 530 nm/λEm = 590 nm.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service