Skip to Content
MilliporeSigma
All Photos(1)

Documents

BI0221

Sigma-Aldrich

Capping Reagent A THF/Lutidine/Acetic Anhydride (8/1/1)

Novabiochem®

Synonym(s):

dna

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116006

Quality Level

product line

Novabiochem®

form

liquid

reaction suitability

reaction type: Acetylations
reagent type: derivatization reagent

transition temp

flash point 4 °C

shipped in

ambient

storage temp.

room temp

Application

BI0221-0450 is packaged in 28/405 glass bottles
BI0221-0950 is packaged in 38/430 glass bottles
BI0221-0960 is packaged in 28/405 glass bottles
BI0221-2500 is packaged in 38/430 glass bottles
BI0221-4000 is packaged in 38/430 glass bottles
DNA Reagents Label Change Explanation
Novabiochem®

Analysis Note

Assay (2,6-lutidine)9.0-11.0% v/vAssay (acetic anhydride)9.0-11.0% v/vAssay (tetrahydrofuran)79.0-81.0% v/vColor (APHA) at time of manufacture50APHA maxFormClear liquidWater (H2O)300ppm max

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

39.2 °F - closed cup

Flash Point(C)

4 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service