Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

MAK348

Sigma-Aldrich

Gelatinase (Gelatin Degradation/Zymography) Assay Kit (Fluorometric)

sufficient for 100 fluorometric tests

Synonym(s):

Fluorometric Gelatinase Activity Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.84

usage

sufficient for 100 fluorometric tests

detection method

fluorometric

storage temp.

−20°C

General description

Gelatinases are a type of matrix zinc-dependent metalloproteases (MMPs) that degrade gelatins and a variety of other extracellular matrix proteins. These enzymes are synthesized as latent zymogens that are activated by proteolysis and inhibited by tissue inhibitors of metalloproteases (TIMPs). Two mammalian gelatinases, Gelatinase A (MMP-2) and Gelatinase B (MMP-9), are critical for basement membrane degradation and are highly upregulated in variety of tumor cells.

Features and Benefits

Compatible with high-throughput handling systems.

Suitability

Suitable for the detection of gelatinase activity in biological samples such as recombinant protein, tissue, cell lysates, etc

Principle

The Gelatinase Activity Assay Kit utilizes a hybrid approach for the detection of gelatinase activity by employing a highly quenched gelatin substrate which upon cleavage by a suitable gelatinase releases a fluorophore, which can be easily quantified using a conventional microplate reader. This method is substrate-specific, simple, fast, high-throughput adaptable and amenable to the sensitive detection of gelatinase activity (as low as 0.06 mCDU).

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

10 - Combustible liquids

Flash Point(F)

188.6 °F

Flash Point(C)

87 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Helena Choltus et al.
International journal of molecular sciences, 22(15) (2021-08-08)
Maternal smoking is a risk factor of preterm prelabor rupture of the fetal membranes (pPROM), which is responsible for 30% of preterm births worldwide. Cigarettes induce oxidative stress and inflammation, mechanisms both implicated in fetal membranes (FM) weakening. We hypothesized

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service