Skip to Content
MilliporeSigma
All Photos(1)

Documents

MAK328

Sigma-Aldrich

Arginase Inhibitor Screening Kit

sufficient for 100 colorimetric tests

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503

usage

sufficient for 100 colorimetric tests

detection method

colorimetric

relevant disease(s)

cardiovascular diseases; neurological disorders

General description

Arginase (L-arginine ureohydrolase) is present in mammals and plants. In humans, arginase is expressed predominantly in the liver, and to lesser degrees in breast, kidney, testes, salivary glands, epidermis and erythrocytes. Arginase catalyzes the conversion of arginine to ornithine and urea, important for protection against NH3 toxicity and for cell growth and repair. Excessive arginase activity has been linked to cardiovascular diseases, also contribute to vascular structural problems and neural toxicity. Studies show that arginase inhibitors have been proven to be beneficial in cardiovascular and nervous system diseases. Simple, direct and automation-ready procedures for measuring arginase inhibition are highly desirable in Research and Drug Discovery.

Features and Benefits

Compatible with high-throughput handling systems.

Suitability

Suitable for inhibitor screening and evaluation of Arginase inhibitors.

Principle

The Arginase Inhibitor Screening Kit provides a sensitive and convenient method to screen for arginase inhibitors. The method utilizes a chromogen that forms a colored complex specifically with urea produced in the arginase reaction. The intensity of the color is directly proportional to the arginase activity in the sample. Percent inhibition of a test compound can be determined by comparing the color intensity of the reaction preincubated with the test compound with the color intensity of an untreated control reaction.

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1 - Met. Corr. 1 - Repr. 1B - Skin Corr. 1A

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service