Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

MAK155

Sigma-Aldrich

Fluorometric Sphingomyelinase Assay Kit

sufficient for 200 fluorometric tests (red fluorescence)

Synonym(s):

Neutral Sphingomyelinase Assay Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

usage

sufficient for 200 fluorometric tests (red fluorescence)

detection method

fluorometric

relevant disease(s)

cancer; rheumatological diseases

storage temp.

−20°C

General description

Sphingomyelinase (SMase), or sphingomyelin phosphodiesterase, catalyzes the hydrolytic cleavage of sphingolipid to phosphocholine and ceramide, a second messenger involved in multiple cellular responses including the cellular response to stress. There are at least five types of SMases identified, which are classified according to their cation dependence, cellular location, and pH optima for activity, and include the acidic and neutral sphingomyelinase. Neutral SMases, along with the acidic SMases, are thought to be major contributors to the production of ceramide in response to cellular stress. Mutations in is associated with metabolic disorders like Niemann-Pick disease B.

Suitability

This kit is suitable for the detection of sphingomyelin in a variety of samples such as blood and cell extracts.

Principle

The Sphingomyelinase Assay Kit provides a simple and direct procedure for measuring neutral SMase activity in a variety of samples such as blood or cell extracts, or for screening SMase activators and inhibitors. SMase activity is determined by a coupled enzyme assay, which results in a fluorescent product (λex =540/λem = 590 nm) proportional to the SMase activity present.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Secretory sphingomyelinase in health and disease.
Kornhuber J, et al.
Biological Chemistry, 396(6-7), 707-736 (2015)
Sphingomyelinases: enzymology and membrane activity.
Go?i F M and Alonso A
Febs Letters, 531(1), 38-46 (2002)
Sphingomyelinases: their regulation and roles in cardiovascular pathophysiology.
Pavoine C and Pecker F
Cardiovascular Research, 82(2), 175-183 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service