Skip to Content
MilliporeSigma
All Photos(3)

Documents

MAK054

Sigma-Aldrich

α-Ketoglutarate Assay Kit

sufficient for 100 colorimetric or fluorometric tests

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

usage

sufficient for 100 colorimetric or fluorometric tests

detection method

colorimetric
fluorometric

relevant disease(s)

cancer

storage temp.

−20°C

General description

α-Ketoglutarate (α-KG) is a key intermediate in the Tricarboxylic Acid (TCA) Cycle, formed by the oxidative decarboylation of isocitrate by isocitrate dehydrogenase. The synthesis of α-KG from glutamate by glutamate dehydrogenase is a key TCA cycle anaplerotic reaction. α-KG is an important nitrogen transporter. This α-Ketoglutarate Assay Kit provides a simple, sensitive and rapid means for quantifying α-KG in a variety of samples.

Application

α-Ketoglutarate Assay Kit has been used to determine the activity of α-ketoglutarate.

Suitability

Suitable for the measurement of a-Ketoglutarate in a variety of biological samples including cell and tissue lysates

Principle

α-KG concentration is determined by a coupled enzyme assay, which results in a colorimetric (570 nm)/ fluorometric (λex = 535/λem = 587 nm) product, proportional to the α-KG present. Typical sensitivities of detection for this kit are 0.01 to 10 nmoles.

related product

Product No.
Description
Pricing

replaced by

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

Flash Point(F)

188.6 °F - closed cup

Flash Point(C)

87 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qun Zhou et al.
Bioscience reports, 39(4) (2019-03-25)
Long intergenic non-coding RNA p21 (lincRNA-p21) is down-regulated in some solid tumors. Glutamine catabolism plays an important role in cancer development. However, the role of lincRNA-p21 and its association with glutamine catabolism remain unknown in bladder cancer (BC). In the
Phoebe J Benson et al.
Frontiers in microbiology, 7, 475-475 (2016-04-20)
Interest in the production of carbon commodities from photosynthetically fixed CO2 has focused attention on cyanobacteria as a target for metabolic engineering and pathway investigation. We investigated the redirection of carbon flux in the model cyanobacterial species, Synechococcus elongatus PCC
Gunars Osis et al.
Physiological reports, 4(8) (2016-04-28)
The bicarbonate transporter, NBCe1 (SLC4A4), is necessary for at least two components of the proximal tubule contribution to acid-base homeostasis, filtered bicarbonate reabsorption, and ammonia metabolism. This study's purpose was to determine NBCe1's role in a third component of acid-base
Sulagna Bhattacharya et al.
EBioMedicine, 68, 103418-103418 (2021-06-09)
We have previously described an evolutionarily selected Tibetan prolyl hydroxylase-2 (PHD2D4E;C127S) variant that degrades the hypoxia-inducible factor (HIFα) more efficiently and protects these highlanders from hypoxia-triggered elevation in haemoglobin concentration. High altitude is known to cause acute mountain sickness (AMS)
Mark A White et al.
Molecular cancer research : MCR, 15(8), 1017-1028 (2017-05-17)
Despite the known importance of androgen receptor (AR) signaling in prostate cancer, the processes downstream of AR that drive disease development and progression remain poorly understood. This knowledge gap has thus limited the ability to treat cancer. Here, it is

Articles

Sigma article discusses tumor cell metabolic pathways, focusing on aerobic glycolysis and mitochondrial activity.

Protocols

We describe here a rapid and sensitive method to separate and measure D-2-OHG and L-2-OHG enantiomers using high-resolution mass spectrometry (HRMS) detection.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service