Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MAK333

Sigma-Aldrich

Citrate Assay Kit

sufficient for 100 colorimetric or fluorometric tests

Synonym(s):

Citric Acid Test Kit

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
$637.00

$637.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 KIT
$637.00

About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

$637.00


Please contact Customer Service for Availability

Request a Bulk Order

usage

sufficient for 100 colorimetric or fluorometric tests

input

culture(s)
plasma
tissue
serum
urine

detection method

colorimetric
fluorometric

relevant disease(s)

cancer

storage temp.

−20°C

General description

Citrate is an intermediate in the citric acid cycle and is involved in fatty acid synthesis. The Citrate Assay Kit provides a simple, and automation-ready procedure for measuring citrate concentration. Citrate is converted into pyruvate which is then oxidized with the conversion of the dye into a colored and fluorescent form. The color intensity at 570 nm or fluorescence intensity at λex/em = 530/585 nm is directly proportional to the citrate concentration in the sample. Linear detection range: 4 to 400 μM citrate for colorimetric assays and 0.5 to 40 μM for fluorimetric assays.

Application

The Citrate Assay Kit may be used for:
  • Cancer Research
  • Drug Discovery
  • Metabolic Disorders Research

Features and Benefits

Efficient and Precise: Detect Citrate levels ranging from 4 to 400 μM (colorimetric) and 0.5 to 40 μM (fluorimetric) in a 15 minute room temperature reaction, providing both convenience and sensitivity.

Compatibility with High-Throughput Systems: Easily incorporate our kit into high-throughput handling systems, ensuring smooth and accurate processing, enhancing efficiency in your laboratory workflow.

Suitability

Suitable for citrate determination in biological samples (e.g. plasma, serum, urine, tissue and culture media.)

Principle

In this kit, citrate is converted into pyruvate which is then oxidized with the conversion of the dye into a colored and fluorescent form. The color intensity at 570 nm or fluorescence intensity at λex/em = 530/585 nm is directly proportional to the citrate concentration in the sample. Linear detection range: 4 to 400 μM citrate for colorimetric assays and 0.5 to 40 μM for fluorimetric assays.

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Storage Class Code

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service