Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

MAK033

Sigma-Aldrich

ADP Colorimetric/Fluorometric Assay Kit

sufficient for 100 colorimetric or fluorometric tests

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.84

usage

sufficient for 100 colorimetric or fluorometric tests

detection method

colorimetric
fluorometric

storage temp.

−20°C

General description

The new ADP Assay Kit, MAK518, is now available! Adenosine diphosphate (ADP) is a nucleoside diphosphate that plays a critical role in energy transfer reactions. ADP is produced from adenosine triphosphate (ATP) via the action of ATPases. ADP also plays a critical role in platelet function. ADP, stored in platelet-dense granules, is released upon platelet activation where it acts on purinergic receptors to mediate intracellular signaling and platelet aggregation. ADP is a key agonist involved in thrombosis and hemostasis.

Features and Benefits

Compatible with high-throughput handling systems.

Suitability

Suitable for the measuerement of ADP in a variety of biological samples including cell and tissue lysates

Principle

In this assay, ADP (adenosine diphosphate) concentration is determined by a coupled enzyme reaction, which results in a colorimetric (570 nm)/fluorometric (λex = 535/λem = 587 nm) product, proportional to the ADP present.

replaced by

Product No.
Description
Pricing

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

Flash Point(F)

188.6 °F - closed cup

Flash Point(C)

87 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Multinuclear magnetic resonance spectroscopy studies of brain purines in major depression.
Renshaw P F, et al.
The American Journal of Psychiatry, 158(12), 2048-2055 (2001)
Molecular identification and characterization of the platelet ADP receptor targeted by thienopyridine antithrombotic drugs.
Foster C J, et al.
The Journal of Clinical Investigation, 107(12), 1591-1598 (2001)
Ana Luisa Palacios-Acedo et al.
Frontiers in oncology, 11, 704945-704945 (2021-10-01)
Platelet function can be modified by cancer cells to support tumor growth, causing alterations in the delicate hemostatic equilibrium. Cancer-cell and platelet interactions are one of the main pillars of Trousseau's syndrome: a paraneoplastic syndrome with recurring and migrating episodes
ADP is not an agonist at P2X1 receptors: evidence for separate receptors stimulated by ATP and ADP on human platelets.
Mahaut-Smith M P, et al.
British Journal of Pharmacology, 131(1), 108-114 (2000)
Sophie Hallakou-Bozec et al.
PloS one, 16(2), e0241651-e0241651 (2021-02-20)
Pancreatic islet β-cell dysfunction is characterized by defective glucose-stimulated insulin secretion (GSIS) and is a predominant component of the pathophysiology of diabetes. Imeglimin, a novel first-in-class small molecule tetrahydrotriazine drug candidate, improves glycemia and GSIS in preclinical models and clinical

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service