Skip to Content
MilliporeSigma
All Photos(3)

Documents

MAK019

Sigma-Aldrich

Maltose Assay Kit

sufficient for 100 colorimetric or fluorometric tests

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

usage

sufficient for 100 colorimetric or fluorometric tests

application(s)

cosmetics
food and beverages

detection method

colorimetric
fluorometric

relevant disease(s)

endocrinological disorders, diabetes

storage temp.

−20°C

General description

The new Maltose Assay Kit, MAK513, is now available! Maltose is a disaccharide containing two glucose molecules with an a(1→4) glycosidic linkage. Maltose can be derived from starch in food through the action of amylase. Maltose can be found in many food products, including beer, cereals, and pasta.

Suitability

Suitable for the direct detection of maltose in a variety of samples, including serum, plasma, food, or growth media.

Principle

The Maltose Assay kit provides a simple and direct procedure for measuring maltose in a variety of samples, including serum, plasma, food, or growth media. In this kit, maltose is converted to two glucose units via α-D-Glucosidase. Glucose is further oxidized, resulting in a colorimetric (570 nm)/fluorometric (λex = 535/λem = 587 nm) product, proportional to the maltose present.

replaced by

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3 - Resp. Sens. 1

Storage Class Code

10 - Combustible liquids

Flash Point(F)

188.6 °F - closed cup

Flash Point(C)

87 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiao-Mei Zhu et al.
Royal Society open science, 6(4), 190418-190418 (2019-06-12)
The production of secondary metabolites, while important for bioengineering purposes, presents a paradox in itself. Though widely existing in plants and bacteria, they have no definite physiological roles. Yet in both native habitats and laboratories, their production appears robust and
Mamta Rai et al.
Cell metabolism, 33(6), 1137-1154 (2021-03-28)
Neurodegeneration in the central nervous system (CNS) is a defining feature of organismal aging that is influenced by peripheral tissues. Clinical observations indicate that skeletal muscle influences CNS aging, but the underlying muscle-to-brain signaling remains unexplored. In Drosophila, we find

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service