Skip to Content
MilliporeSigma
All Photos(1)

Documents

M071000

Sigma-Aldrich

5-(Ethylthio)-1H-tetrazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H6N4S
CAS Number:
Molecular Weight:
130.17
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:

type

for DNA synthesis

product line

Proligo Reagents

Assay

≥99.0% (reversed phase HPLC)

impurities

≤300 ppm water content (Karl Fischer)

color

solid white

mp

84-90 °C (lit.)
86-88 °C

suitability

conforms to structure for H-NMR

SMILES string

CCSc1nnn[nH]1

InChI

1S/C3H6N4S/c1-2-8-3-4-6-7-5-3/h2H2,1H3,(H,4,5,6,7)

InChI key

GONFBOIJNUKKST-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

4.1A - Other explosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service