Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

J4024

Sigma-Aldrich

JHW 007 hydrochloride

≥98% (HPLC), solid

Synonym(s):

(3-endo)-3-[Bis(4-fluorophenyl)methoxy]-8-butyl-8-azabicyclo[3.2.1]octane hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H29F2NO · HCl
CAS Number:
Molecular Weight:
421.95
MDL number:
UNSPSC Code:
12352200

Assay

≥98% (HPLC)

form

solid

color

white

solubility

H2O: ~10 mg/mL

storage temp.

2-8°C

SMILES string

CCCCN1C2CCC1CC(C2)OC(c3ccc(F)cc3)c4ccc(F)cc4

Biochem/physiol Actions

Selective antagonist of the cocaine site on dopamine transporter.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G E Agoston et al.
Journal of medicinal chemistry, 40(26), 4329-4339 (1998-01-22)
A series of N-substituted 3 alpha-[bis(4'-fluorophenyl)methoxy]tropane analogues has been prepared that function as dopamine uptake inhibitors. The N-methylated analogue of this series had a significantly higher affinity for the dopamine transporter than the parent compound, N-methyl-3 alpha- (diphenylmethoxy)tropane (benztropine, Cogentin).
Rajeev I Desai et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 25(8), 1889-1893 (2005-02-25)
There is a large unmet medical need for cocaine addiction treatments. Studies have indicated that the dopamine transporter (DAT) is the primary biological target of cocaine, and most drugs that have DAT affinity have behavioral effects like those of cocaine.
Sangeeta Raje et al.
The Journal of pharmacology and experimental therapeutics, 307(2), 801-808 (2003-09-11)
The N-substituted 3alpha-[bis(4'-fluorophenyl)methoxy]tropanes (AHN 2-003, AHN 1-055, AHN 2-005, and JHW 007) bind with high affinity to the dopamine transporter and inhibit dopamine uptake more potently than cocaine, but they demonstrate behavioral profiles in animal models of psychostimulant abuse that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service