MilliporeSigma
  • Identification of a dopamine transporter ligand that blocks the stimulant effects of cocaine.
Identification of a dopamine transporter ligand that blocks the stimulant effects of cocaine.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2005-02-25)
Rajeev I Desai, Theresa A Kopajtic, Mikhail Koffarnus, Amy Hauck Newman, Jonathan L Katz
ABSTRACT
There is a large unmet medical need for cocaine addiction treatments. Studies have indicated that the dopamine transporter (DAT) is the primary biological target of cocaine, and most drugs that have DAT affinity have behavioral effects like those of cocaine. However, analogs of benztropine have high DAT affinity and behavioral effects that show varying degrees of similarity to cocaine. We now report the discovery that a benztropine analog, JHW007, with high affinity for the DAT does not have cocaine-like behavioral effects and antagonizes the effects of cocaine. JHW007 occupied the DAT in vivo more slowly than did cocaine and had not reached an apparent plateau up to 270 min after injection. The in vivo binding of cocaine to the DAT suggested rate of DAT occupancy as an important contributor to its behavioral effects, and the slow association with the DAT may provide an explanation for JHW007 being relatively devoid of cocaine-like behavioral effects. The antagonism of cocaine suggests that DAT ligands with reduced cocaine-like activity can function as cocaine antagonists and suggests JHW007 as a lead for discovery of cocaine-abuse pharmacotherapeutics.