Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

I9882

Sigma-Aldrich

1-Iodopropane

~99%

Synonym(s):

Propyl iodide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2I
CAS Number:
Molecular Weight:
169.99
Beilstein:
505937
MDL number:
UNSPSC Code:
12352100

vapor pressure

43 mmHg ( 25 °C)

Assay

~99%

contains

copper as stabilizer

refractive index

n20/D 1.504 (lit.)

bp

101-102 °C (lit.)

mp

−101 °C (lit.)

density

1.743 g/mL at 25 °C (lit.)

SMILES string

CCCI

InChI

1S/C3H7I/c1-2-3-4/h2-3H2,1H3

InChI key

PVWOIHVRPOBWPI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 1 - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 3 - Muta. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

111.2 °F

Flash Point(C)

44 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service