Skip to Content
MilliporeSigma
1-Iodopropane 99%

1-Iodopropane

1-Iodopropane 99%
Synonym(s):
Propyl iodide, 1-Iodopropane
Linear Formula:
CH3CH2CH2I
CAS Number:
Molecular Weight:
169.99

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing