Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

I3263

Sigma-Aldrich

Insulin-like Growth Factor-II fragment 33-40

≥97% (HPLC)

Synonym(s):

Ser-Arg-Val-Ser-Arg-Arg-Ser-Arg

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C38H74N20O12
CAS Number:
Molecular Weight:
1003.12
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥97% (HPLC)

composition

Peptide content, ~65%

UniProt accession no.

storage temp.

−20°C

SMILES string

CC(C)C(NC(=O)C(CCCNC(N)=N)NC(=O)C(N)CO)C(=O)NC(CO)C(=O)NC(CCCNC(N)=N)C(=O)NC(CCCNC(N)=N)C(=O)NC(CO)C(=O)NC(CCCNC(N)=N)C(O)=O

InChI

1S/C38H74N20O12/c1-18(2)26(58-30(65)22(9-5-13-50-37(44)45)52-27(62)19(39)15-59)33(68)57-25(17-61)31(66)54-20(7-3-11-48-35(40)41)28(63)53-21(8-4-12-49-36(42)43)29(64)56-24(16-60)32(67)55-23(34(69)70)10-6-14-51-38(46)47/h18-26,59-61H,3-17,39H2,1-2H3,(H,52,62)(H,53,63)(H,54,66)(H,55,67)(H,56,64)(H,57,68)(H,58,65)(H,69,70)(H4,40,41,48)(H4,42,43,49)(H4,44,45,50)(H4,46,47,51)

InChI key

ZLWOQMRVOZVGEB-UHFFFAOYSA-N

Gene Information

rat ... Npy(24604)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R L Hintz et al.
The Journal of clinical endocrinology and metabolism, 54(2), 442-446 (1982-02-01)
Insulin-like growth factor II (IGF-II) is a human plasma peptide whose sequence is homologous to both insulin-like growth factor I/somatomedin C (IGF-I/SM-C) and human proinsulin in the A and B regions. However, there is no obvious homology in the C

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service