Skip to Content
MilliporeSigma
All Photos(1)

Documents

I0514

Sigma-Aldrich

Diethyl isocyanomethylphosphonate

>97% (GC)

Synonym(s):

Isocyanomethylphosphonic acid diethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CNCH2P(O)(OC2H5)2
CAS Number:
Molecular Weight:
177.14
Beilstein:
4674920
MDL number:
UNSPSC Code:
12352000

Assay

>97% (GC)

refractive index

n20/D 1.433 (lit.)

bp

84-87 °C/0.1 mmHg (lit.)

density

1.105 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CCOP(=O)(C[N+]#[C-])OCC

InChI

1S/C6H12NO3P/c1-4-9-11(8,6-7-3)10-5-2/h4-6H2,1-2H3

InChI key

ICURVXBGGDVHDT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant for:
  • Copper-catalyzed cycloaddition reactions
  • Preparation of orally bioavailable diamide prodrugs that lower glucose in diabetic animals through inhibition of fructose-1,6-bisphosphatase
  • Synthesis of sterol analogs as antifungal agents against plant pathogens
  • Asymmetric synthesis of steroidal 2,5-diketopiperazines prepared via isocyanide chemistry based on stereoselective Ugi four component condensation and subsequent cyclocondensation reactions
  • The Passerini reaction
Precursor for synthesis of phosphonic acid analogs of α-amino acids.

replaced by

Product No.
Description
Pricing

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rachon, J.
Synthesis, 219-219 (1984)
Schollkopf, U. and Wintel, T.
Synthesis, 1033-1033 (1984)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service