Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

359866

Sigma-Aldrich

Diethyl isocyanomethylphosphonate

97%

Synonym(s):

Isocyanomethylphosphonic acid diethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CNCH2P(O)(OC2H5)2
CAS Number:
Molecular Weight:
177.14
Beilstein:
4674920
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22
form:
liquid
Assay:
97%

Assay

97%

form

liquid

reaction suitability

reaction type: C-C Bond Formation

refractive index

n20/D 1.433 (lit.)

bp

84-87 °C/0.1 mmHg (lit.)

density

1.105 g/mL at 25 °C (lit.)

functional group

amine
isonitrile
phosphonate

storage temp.

2-8°C

SMILES string

CCOP(=O)(C[N+]#[C-])OCC

InChI

1S/C6H12NO3P/c1-4-9-11(8,6-7-3)10-5-2/h4-6H2,1-2H3

InChI key

ICURVXBGGDVHDT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant for:
  • Copper-catalyzed cycloaddition reactions
  • Preparation of orally bioavailable diamide prodrugs that lower glucose in diabetic animals through inhibition of fructose-1,6-bisphosphatase
  • Synthesis of sterol analogs as antifungal agents against plant pathogens
  • Asymmetric synthesis of steroidal 2,5-diketopiperazines prepared via isocyanide chemistry based on stereoselective Ugi four component condensation and subsequent cyclocondensation reactions
  • The Passerini reaction

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service