Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

HPA002656

Sigma-Aldrich

Anti-IFIH1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Helicard antibody produced in rabbit, Anti-Helicase with 2 CARD domains antibody produced in rabbit, Anti-Interferon-induced helicase C domain-containing protein 1 antibody produced in rabbit, Anti-Interferon-induced with helicase C domain protein 1 antibody produced in rabbit, Anti-MDA-5 antibody produced in rabbit, Anti-Melanoma differentiation-associated protein 5 antibody produced in rabbit, Anti-RNA helicase-DEAD box protein 1 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$757.00

$757.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μL
$757.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

$757.00


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

TIRMIDAYTHLETFYNEEKDKKFAVIEDDSDEGGDDEYCDGDEDEDDLKKPLKLDETDRFLMTLFFENNKMLKRLAENPEYENEKLTKLRNTIMEQYTRTEESARGIIFTKTRQSAYALSQWITENEKFAEVGVKAHHLIGAGHSSEFKP

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... IFIH1(64135)

General description

IFIH1 (interferon induced with helicase C domain 1) is mainly involved in the innate immune response triggered by viral infection. It is also known as the melanoma differentiation-associated gene-5 (MDA-5) and encodes a cytoplasmic receptor of the pattern-recognition receptors (PRRs) family.

Immunogen

Interferon-induced helicase C domain-containing protein 1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

IFIH1 (interferon induced with helicase C domain 1) is actively involved in initialization of innate immunity response signaling cascade. It initiates the transcription of innate response genes, such as α/β interferon (IFN α/β) proinflammatory cytokines. Interferons protect uninfected cells and induce apoptosis in infected cells. During viral infection, annealing of complementary RNAs produce long length double-stranded RNAs which helps to activate IFIH1. It acts as a candidate gene for the type 1 diabetes mellitus (T1DM). IFIH1 has developmental impact on the autoimmune disease. Mutation in IFIH1 causes a rare, genetically early-onset progressive encephalopathy disorder named as Aicardi-Goutières syndrome (AGS).

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST83080

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ana Paula Bouças et al.
Arquivos brasileiros de endocrinologia e metabologia, 57(9), 667-676 (2014-01-10)
Type 1 diabetes mellitus (T1DM) is a chronic, progressive, autoimmune disease characterized by metabolic decompensation frequently leading to dehydration and ketoacidosis. Viral pathogens seem to play a major role in triggering the autoimmune destruction that leads to the development of
Hirotsugu Oda et al.
American journal of human genetics, 95(1), 121-125 (2014-07-06)
Aicardi-Goutières syndrome (AGS) is a rare, genetically determined early-onset progressive encephalopathy. To date, mutations in six genes have been identified as etiologic for AGS. Our Japanese nationwide AGS survey identified six AGS-affected individuals without a molecular diagnosis; we performed whole-exome
Andreas Pichlmair et al.
Journal of virology, 83(20), 10761-10769 (2009-08-07)
Recognition of virus presence via RIG-I (retinoic acid inducible gene I) and/or MDA5 (melanoma differentiation-associated protein 5) initiates a signaling cascade that culminates in transcription of innate response genes such as those encoding the alpha/beta interferon (IFN-alpha/beta) cytokines. It is

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service