Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

H0417

Sigma-Aldrich

Heparin Cofactor II from human plasma

aqueous glycerol solution, ≥90% (SDS-PAGE)

Synonym(s):

HCF2, HCII, SERPIND1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$851.00

$851.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
$851.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.61

$851.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

human plasma

Quality Level

Assay

≥90% (SDS-PAGE)

form

aqueous glycerol solution

mol wt

~66 kDa
~66 kDa

NCBI accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... SERPIND1(3053)

Biochem/physiol Actions

Heparin cofactor II (HCII), a member of the serpin superfamily, is a serine protease inhibitor that rapidly inactivates thrombin in the presence of dermatan sulfate, heparan sulfate, or heparin. In addition to its role in coagulation regulation, HCII may also participate in inflammation, atherosclerosis, and wound repair. For example, leukocyte-mediated degradation of HCII releases peptides that are chemotactic for neutrophils and monocytes which may promote acute inflammation and wound repair. It has also been reported that HCII and dermatan sulfate levels are elevated in pregnancy, suggesting that HCII may be protective against thrombosis during pregnancy.
Serine protease that inhibits thrombin, α-chymotrypsin, neutrophil cathepsin G, and Streptomyces griseus protease B.

Unit Definition

HCII is tested for activity using a Thrombin inhibition assay. One unit will inhibit one NIH unit of Thrombin.

Physical form

50% aqueous glycerol solution (vol/vol)

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. Will Heparin cofactor II (Sigma H0417) stop blood from coagulating?

    1 answer
    1. Heparin Cofactor II (HCII) is a member of the serpin superfamily and acts as a serine protease inhibitor. It rapidly inactivates thrombin in the presence of dermatan sulfate, heparan sulfate, or heparin. However, it should be noted that Heparin Cofactor II itself is not a proper anticoagulant. When used alone, it will not prevent blood from coagulating. For anticoagulant purposes, substances such as Heparin sodium salt, EDTA, or Sodium Citrate are commonly used in research and clinical laboratories.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service