Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GW22561

Sigma-Aldrich

Anti-SNX2 antibody produced in chicken

affinity isolated antibody, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

chicken

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

mouse, human, rat

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SNX2(6643)

General description

SNX2 (sorting nexin 2) is a member of cytoplasmic and membrane proteins called SNXs. This protein is expressed in thyroid in the columnar and active follicular cells. It contains the PX (phox homology) domain, found in all SNX members, which binds to phosphatidylinositol 3-phosphate (PI3P). It also contains a BAR domain adjacent to its PX domain, towards the C-terminal. It is thought to form a dimer with other SNX member proteins, which in turn make a part of the heteropentameric complex called retromer. SNX2 also contains a coiled-coil domain in its C-terminal. It is a homologue of vacuolar protein-sorting (Vps)5p found in yeast. It has a molecular weight of ~68kDa.

Immunogen

Immunogen Sequence: GI # 4507141, sequence 1-519
Recombinant sorting nexin 2

Application

Anti-SNX2 antibody produced in chicken is suitable for western blotting analysis at a dilution of 1:500, for tissue or cell staining at a dilution of 1:200.

Biochem/physiol Actions

Sorting nexin-2 (SNX2) acts as a novel marker for active thyrocytes and reflects the endosomal trafficking activity in these cells. It forms a component of the mammalian retromer complex that functions in the retrieval of the cation-independent mannose 6-phosphate receptor from endosomes to the trans-Golgi network. It is involved, though not necessary, in the lysosomal sorting of EGFR (epidermal growth factor receptor), which is internalized. Mouse embryos with inactivated SNX2 gene die mid-gestation. This study suggests that SNX2 is essential during developmental processes.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maki Kanzawa et al.
Applied immunohistochemistry & molecular morphology : AIMM, 22(4), 302-307 (2013-03-28)
Sorting nexins (SNXs) are a large, diverse group of cytoplasmic and membrane-associated proteins that function in a variety of cellular processes, including endocytosis, protein trafficking, and the retrieval of transmembrane proteins. In this study, we demonstrated that SNX2 is expressed
Jez G Carlton et al.
Journal of cell science, 118(Pt 19), 4527-4539 (2005-09-24)
Sorting nexins are a large family of phox-homology-domain-containing proteins that have been implicated in the control of endosomal sorting. Sorting nexin-1 is a component of the mammalian retromer complex that regulates retrieval of the cation-independent mannose 6-phosphate receptor from endosomes
Juan S Bonifacino et al.
Current opinion in cell biology, 20(4), 427-436 (2008-05-13)
The retromer is a heteropentameric complex that associates with the cytosolic face of endosomes and mediates retrograde transport of transmembrane cargo from endosomes to the trans-Golgi network. The mammalian retromer complex comprises a sorting nexin dimer composed of a still
Anuradha Gullapalli et al.
Molecular biology of the cell, 15(5), 2143-2155 (2004-02-24)
Sorting nexin 1 (SNX1) and SNX2, homologues of the yeast vacuolar protein-sorting (Vps)5p, contain a phospholipid-binding motif termed the phox homology (PX) domain and a carboxyl terminal coiled-coil region. A role for SNX1 in trafficking of cell surface receptors from

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service