MilliporeSigma
  • Sorting Nexin 2 (SNX2): a potential marker of active thyrocytes in normal and hyperfunctioning thyroid disorders.
Sorting Nexin 2 (SNX2): a potential marker of active thyrocytes in normal and hyperfunctioning thyroid disorders.
Applied immunohistochemistry & molecular morphology : AIMM (2013-03-28)
Maki Kanzawa, Shigeo Hara, Shuho Semba, Hiroshi Yokozaki, Mitsuyoshi Hirokawa, Tomoo Itoh
ABSTRACT
Sorting nexins (SNXs) are a large, diverse group of cytoplasmic and membrane-associated proteins that function in a variety of cellular processes, including endocytosis, protein trafficking, and the retrieval of transmembrane proteins. In this study, we demonstrated that SNX2 is expressed in columnar and active thyroid follicular cells but not in flattened inactive thyrocytes. Morphometric analysis revealed a significant correlation between SNX2 positivity and columnar cell morphology. Immunohistochemical staining of serial sections of the thyroid tissue indicated that SNX2 localization was similar to sortilin, a protein expressed by active thyrocytes. Expression of SNX2 in thyrocytes is particularly marked and extensive in most hyperstimulated thyroid disorders, including Graves disease (diffusely SNX2 positive in 73.3% patients) and functioning nodules (93.8% patients). SNX2 immunolocalization in hyperstimulated follicular epithelial cells was specific among the SNXs family members examined. These results support the utility of SNX2 as a novel marker of active thyrocytes and reflect the endosomal trafficking activity in these cells.