GERPN1607
Megaprime™ DNA Labelling System, dCTP
Cytiva RPN1607, sufficient for 60 reactions
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106000
NACRES:
NA.31
Recommended Products
usage
sufficient for 60 reactions
manufacturer/tradename
Cytiva RPN1607
shipped in
dry ice
storage temp.
−20°C
General description
Megaprime™ DNA Labeling System, dCTP (for use with labeled dCTP).
Application
Megaprime™ DNA Labeling Systems are used for rapid, random-prime labeling from as little as 25 ng substrate DNA. Each system includes the following reagents, sufficient for 30 or 60 reactions (depending on product ordered): primer solution, labeling buffer, reaction buffer, enzyme, standard DNA solution, and carrier DNA solution.
Features and Benefits
- For labeling of DNA probes to specific activities > 1 × 109 dpm/μg in 10 min.
- Features cloned Klenow fragment and random nonamers in the random-prime labeling technique.
- No need to remove unincorporated nucleotides before hybridization.
- Protocols for labeling directly from Low melting point Agarose.
Storage and Stability
Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Analysis Note
To view the Certificate of Analysis for this product, please visit www.cytiva.com.
Legal Information
Megaprime is a trademark of Cytiva
Storage Class Code
12 - Non Combustible Liquids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service