Skip to Content
MilliporeSigma
All Photos(1)

Documents

10814270001

Roche

Primer for cDNA Synthesis

lyophilized, pkg of 40 μg

Synonym(s):

oligo-p(dt)15

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106300

form

lyophilized

Quality Level

packaging

pkg of 40 μg

manufacturer/tradename

Roche

storage temp.

−20°C (−15°C to −25°C)

General description

This primer consists of a 15-mer oligonucleotide that shows thymidine (T) at each individual position. The oligonucleotides are HPLC purified, desalted, and supplied as a lyophilizate of the Li-salt. The p(dT)15 primer is not phosphorylated at the 5′ end.Oligo(dT)N generates full-length cDNAs. If template contains oligo(A) stretches internally, the primer may bind these, causing mis priming and transcripts that are not full-length.

Composition: p(dT)15[ 15-mer oligonucleotide containing only deoxythymidine (dT) residues].

Phosphorylation: The p(dT)15 primer is not phosphorylated at the 5′ end.

Application

Primer for cDNA synthesis may be used in:
  • RT-PCR
  • Generation of cDNA libraries
  • Synthesis of first-strand cDNA

Unit Definition

8 nmol ξ 1 A260 unit

Other Notes

For life science research only. Not for use in diagnostic procedures.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Khalil Farhat et al.
Frontiers in immunology, 9, 2175-2175 (2018-10-20)
The precise impact of the somatotrope axis upon the immune system is still highly debated. We have previously shown that mice with generalized ablation of growth hormone (GH) releasing hormone (GHRH) gene (Ghrh-/-) have normal thymus and T-cell development, but
Agnes Bonnet et al.
BMC genomics, 12, 417-417 (2011-08-20)
Successful achievement of early folliculogenesis is crucial for female reproductive function. The process is finely regulated by cell-cell interactions and by the coordinated expression of genes in both the oocyte and in granulosa cells. Despite many studies, little is known
Bingshe Han et al.
PloS one, 7(5), e36941-e36941 (2012-06-01)
Our previous study shows that inhibiting activator protein one (AP1) transcription factor function in murine epidermis, using dominant-negative c-jun (TAM67), increases cell proliferation and delays differentiation. To understand the mechanism of action, we compare TAM67 impact in mouse epidermis and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service