Skip to Content
MilliporeSigma
All Photos(1)

Documents

GE28-9857-99

Streptavidin Mag Sepharose

Cytiva 28-9857-99, pack of 5 × 1 mL

Synonym(s):

Streptavidin Magnetic Beads

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106500
NACRES:
NA.56

packaging

pack of 5 × 1 mL

manufacturer/tradename

Cytiva 28-9857-99

storage condition

(20% Ethanol, 10% medium slurry)

capacity

>300 μg binding capacity(biotinulated BSA/ml of medium slurry)

storage temp.

2-8°C

General description

Streptavidin Mag Sepharose is a magnetic bead for simple and efficient enrichment of target proteins by immunoprecipitation and purification of biotinylated biomolecules. Streptavidin Mag Sepharose utilizes the strong interaction between biotin and streptavidin ligand, which is immobilized on magnetic beads. Magnetic beads simplify sample handling in small-scale purifications.

Application

Streptavidin Mag Sepharose is a magnetic bead for simple and efficient enrichment of target Proteins by immunoprecipitation and purification of biotinylated biomolecules. Streptavidin Mag Sepharose utilizes the strong interaction between biotin and streptavidin ligand, which is immobilized on magnetic beads. Magnetic beads simplify sample handling in small-scale purifications.

The beads are available in two pack sizes: 2 × 1 mL 10% medium slurry and 5 × 1 mL 10% medium slurry. A milliliter of 10% medium slurry is the same as 100 μL sedimented medium and it is sufficient for 20 purification runs according to the recommended immunoprecipitation protocol.

Features and Benefits

  • High-capacity small-scale purification of Proteins from cell lysates and biological fluids
  • High purity and yield
  • Easy parallel screening of Proteins with high repeatability
  • Simple enrichment of Proteins from small or large sample volumes (Low μL to high mL scale.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

Sepharose is a trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Storage Class Code

3 - Flammable liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service