Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GE17-5443-10

Capto DEAE

Cytiva 17-5443-10, pack of 25 mL

Sign Into View Organizational & Contract Pricing

Select a Size

1 PKG
$165.00

$165.00


Please contact Customer Service for Availability


Select a Size

Change View
1 PKG
$165.00

About This Item

UNSPSC Code:
47101511
NACRES:
NA.56

$165.00


Please contact Customer Service for Availability

ligand

diethylaminoethyl

description

Ion Exchanger Type (value)

packaging

pack of 25 mL

manufacturer/tradename

Cytiva 17-5443-10

matrix

highly cross-linked agarose with dextran surface extender

average diameter

90 μm (d50v)

cleaning in place

2-14

working range

2-12

suitability

suitable for bioprocess medium

Related Categories

General description

Capto DEAE is a weak anion exchanger for large-scale manufacturers. Meets demands for fast, cost-effective capture and intermediate protein purification.

Capto DEAE is designed for capture and intermediate purification of proteins from large feed volumes. It is composed of a rigid, high-flow agarose matrix modified with dextran surface extenders and a weak diethylaminoethyl (DEAE) anion exchanger.

The combination of high volume throughput and high dynamic binding capacity for Capto DEAE raises the productivity in industrial downstream processes. Capto DEAE withstands effective and rigorous CIP procedure. The chemical stability supports a long resin lifetime.

Features and Benefits

  • Weak anion exchanger for high-productivity capture and intermediate purification when high volume throughput is essential.
  • High dynamic binding capacity at high flow raises productivity
  • High-volume throughput cuts process times
  • Cost-effective processing with smaller unit operations
  • Resin fulfills industrial demands for security of supply, robust performance, and regulatory support.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Store at 4 to 30 °C (20% Ethanol)

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

Capto is a trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Storage Class Code

3 - Flammable liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service