Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GE10600042

Amersham Protran® Western blotting membranes, nitrocellulose

pore size 0.45 μm, sheet W × L 200 mm × 200 mm, pkg of 10 ea

Sign Into View Organizational & Contract Pricing

Select a Size

10 EA
$426.00

$426.00


Please contact Customer Service for Availability


Select a Size

Change View
10 EA
$426.00

About This Item

UNSPSC Code:
41105339
NACRES:
NB.22

$426.00


Please contact Customer Service for Availability

packaging

pkg of 10 ea

manufacturer/tradename

Cytiva 10600042

sheet W × L

200 mm × 200 mm

pore size

0.45 μm pore size

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Nitrocellulose (NC) membrane for use in Western blot protocols, offering low background and high signal-to-noise ratios. Amersham Protran 0.45 NC is a nitrocellulose membrane with a 0.45 µm pore size for use in Western blot protocols. Amersham Protran nitrocellulose membranes offer excellent sensitivity, resolution, and low background noise, especially when used with Amersham ECL, Amersham ECL Prime, or Amersham ECL Select Western Blotting detection reagents. Protran NC membranes are available in popular precut sizes for a wide range of laboratory applications. This product was formerly called Protran BA85.

Application

Amersham Protran® Western blotting membranes, nitrocellulose is suitable for use in:
  • Western blotting
  • Southern blotting
  • Northern blotting

Features and Benefits

  • Low background and excellent signal-to-noise ratios
  • Compatible with a variety of labelling and detection systems
  • High binding capacity
  • Suitable for proteins with a wide range of molecular weights
  • Ideal tool for your antibody purification and analysis workflow
  • Proprietary nitrocellulose formula ensures a long shelf life of bound proteins

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Legal Information

Amersham is a trademark of Cytiva
Protran is a registered trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Precautionary Statements

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service