Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GE10600002

Amersham Protran® Western blotting membranes, nitrocellulose

pore size 0.45 μm, roll W × L 300 mm × 4 m, pkg of 1 ea

Synonym(s):

Nitrocellulose membranes, Western blot membranes

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$555.00

$555.00


Please contact Customer Service for Availability


Select a Size

Change View
1 EA
$555.00

About This Item

UNSPSC Code:
41105339
NACRES:
NB.22

$555.00


Please contact Customer Service for Availability

packaging

pkg of 1 ea

manufacturer/tradename

Cytiva 10600002

roll W × L

300 mm × 4 m

pore size

0.45 μm pore size

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Amersham Protran 0.45 NC is a nitrocellulose membrane with a 0.45 µm pore size for use in Western blot protocols. Amersham Protran nitrocellulose membranes offer excellent sensitivity, resolution, and low background noise, especially when used with Amersham ECL, Amersham ECL Prime, or Amersham ECL Select Western Blotting detection reagents. Protran NC membranes are available in popular precut sizes for a wide range of laboratory applications. This product was formerly called Protran BA85.

Application

Amersham Protran® Western blotting membranes, nitrocellulose is suitable for use in:
  • Western blotting
  • Southern blotting
  • Northern blotting

Features and Benefits

  • Low background and excellent signal-to-noise ratios
  • Compatible with a variety of labelling and detection systems
  • High binding capacity
  • Suitable for proteins with a wide range of molecular weights
  • Ideal tool for your antibody purification and analysis workflow
  • Proprietary nitrocellulose formula ensures a long shelf life of bound proteins

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Legal Information

Amersham is a trademark of Cytiva
Protran is a registered trademark of Cytiva

Storage Class Code

4.1B - Flammable solid hazardous materials


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Emilia A H Vanni et al.
PLoS pathogens, 16(12), e1009166-e1009166 (2020-12-29)
Herpes simplex virus 1 (HSV-1) infects skin and mucosal epithelial cells and then travels along axons to establish latency in the neurones of sensory ganglia. Although viral gene expression is restricted during latency, the latency-associated transcript (LAT) locus encodes many
Marianna Lucafò et al.
Cancer chemotherapy and pharmacology, 86(3), 361-374 (2020-08-11)
Glucocorticoids (GCs) are commonly used as therapeutic agents for immune-mediated diseases and leukemia. However, considerable inter-individual differences in efficacy have been reported. Several reports indicate that the inhibitor of mTOR rapamycin can reverse GC resistance, but the molecular mechanism involved

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service