Skip to Content
MilliporeSigma
All Photos(1)

Documents

GE10600009

Amersham Protran® Premium Western blotting membranes, nitrocellulose

pore size 0.2 μm, roll W × L 200 mm × 4 m, pkg of 1 ea

Synonym(s):

Nitrocellulose membranes, Western blot membranes

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105339
NACRES:
NB.22

packaging

pkg of 1 ea

manufacturer/tradename

Cytiva 10600009

roll W × L

200 mm × 4 m

pore size

0.2 μm pore size

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Nitrocellulose blotting membrane, provides excellent sensitivity, resolution, and low background in fluorescence Western blotting detection applications.

Application

Amersham Protran® Premium Western blotting membranes, nitrocellulose may be used in:
  • Western blotting
  • Southern blotting
  • Northern blotting

Features and Benefits

  • The membrane is suitable for fluorescence-based immunodetection and multiplexing due to its low auto-fluorescence across a wide range of excitation/emission wavelengths
  • Provides high binding capacity (173 - 203 μg IgG/cm2) of small proteins (< Mr 20 000)
  • Decreases the risk of "blow-through"
  • Precut sizes are available for laboratory application
  • Offers excellent sensitivity, resolution, and low background especially when used with the fluorescence Amersham ECL plex Western blotting detection system

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Legal Information

Amersham is a trademark of Cytiva
Protran is a registered trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Precautionary Statements

Storage Class Code

11 - Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service