Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

G6376

Sigma-Aldrich

β-Glycerophosphate disodium salt hydrate

≤2.4 mol % α-isomer(based on C3H7O6PNa2 With x H2O)

Synonym(s):

BGP, Glycerol 2-phosphate disodium salt hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(HOCH2)2CHOP(O)(ONa)2 · xH2O
CAS Number:
Molecular Weight:
216.04 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:

biological source

synthetic

form

powder

impurities

≤2.4 mol % α-isomer (based on C3H7O6PNa2 With x H2O)

mp

102-104 °C (lit.)

solubility

water: 100 mg/mL, clear, colorless to very faintly yellow

SMILES string

O.[Na+].[Na+].OCC(CO)OP([O-])([O-])=O

InChI

1S/C3H9O6P.2Na.H2O/c4-1-3(2-5)9-10(6,7)8;;;/h3-5H,1-2H2,(H2,6,7,8);;;1H2/q;2*+1;/p-2

InChI key

ROPZSVKNEIIIDE-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

β-Glycerophosphate is a classical serine-threonine phosphatase inhibitor used in kinase reaction buffers. BGP is often used in combination with other phosphatase/protease inhibitors for broad spectrum inhibition. It functions as an organic phosphate donor and has been used in culture media for mesenchymal stem cell differentiation to osteoblast-type cells. BGP is also used to buffer M17 media for Lactococcus culture in recombinant protein expression.
Initiates differentiation of bone-marrow-derived stem cells embedded in a natural collagen/chitosan gel designed for tissue engineering.

comparable product

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anthea Di Rita et al.
International journal of molecular sciences, 21(1) (2020-01-18)
The selective elimination of dysfunctional mitochondria through mitophagy is crucial for preserving mitochondrial quality and cellular homeostasis. The most described mitophagy pathway is regulated by a positive ubiquitylation feedback loop in which the PINK1 (PTEN induced kinase 1) kinase phosphorylates
Lucas R Brun et al.
SpringerPlus, 3, 676-676 (2014-12-19)
The aim of this study was to evaluate the effect of strontium ranelate (SrR) on bone mineral density (BMD) and bone turnover markers after 1 year of treatment. Additionally, the effect of SrR in bisphosphonate-naïve patients (BP-naïve) compared to patients previously
Ya-Ting Deng et al.
PloS one, 10(8), e0136572-e0136572 (2015-09-01)
Echinocystic acid (EA) is a natural triterpone enriched in various herbs and has been used for medicinal purposes in China. In the present study, we systematically examined the effects of EA on ovariectomy-induced osteoporosis in rats for the first time.
Amit Alexander et al.
The Journal of pharmacy and pharmacology, 66(9), 1240-1250 (2014-05-16)
In this study, we have used melphalan (ML) as a model drug, used extensively for the treatment of breast cancer. Due to its remarkable haemolytic activity, clinical application of this drug is limited. We incorporated the two synthesized PEGylated melphalan
Peng Sun et al.
Journal of cellular physiology, 234(7), 10855-10867 (2018-12-12)
The key signaling networks regulating bone marrow mesenchymal stem cells (BMSCs) are poorly defined. Lgr4, which belongs to the leucine-rich repeat-containing G protein-coupled receptor (LGR) family, is widely expressed in multiple tissues from early embryogenesis to adulthood. We investigated whether

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service