Skip to Content
MilliporeSigma
β-Glycerophosphate disodium salt hydrate ≤1.0 mol % L-α-isomer

β-Glycerophosphate disodium salt hydrate

β-Glycerophosphate disodium salt hydrate ≤1.0 mol % L-α-isomer
Synonym(s):
BGP, Glycerol 2-phosphate disodium salt hydrate
Linear Formula:
(HOCH2)2CHOP(O)(ONa)2 · xH2O
CAS Number:
Molecular Weight:
216.04 (anhydrous basis)
EC Number:

Products

Brand
Biological Source
Color
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Research Area
Quality Segment
Technique
Showing 1-2 of 2