Skip to Content
MilliporeSigma
All Photos(1)

Documents

G211050

Sigma-Aldrich

DMT-2′O-TBDMS-rG(ib) Phosphoramidite

configured for (ÄKTA® and OligoPilot®)

Synonym(s):

N-(2-methyl-1-oxopropyl)-5′-O-[bis(4-methoxyphenyl)phenylmethyl]-2′-O-[(1,1-dimethylethyl)dimethylsilyl]-guanosine, 3′-[2-cyanoethyl N,N-bis(1-methylethyl)phosphoramidite]

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C50H68N7O9PSi
CAS Number:
Molecular Weight:
970.18
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.51

biological source

non-animal source (no BSE/TSE risk)

Quality Level

product line

Proligo Reagents

Assay

≥99% (31P-NMR)
≥99.0% (reversed phase HPLC)

form

powder

mol wt

970.18 g/mol

technique(s)

oligo synthesis: suitable

impurities

≤0.3% water content (Karl Fischer)
≤0.5% P(III) Impurities 100-169ppm (31P-NMR)
≤0.5% single Impurity (reversed phase HPLC)
≤3% residual Solvent content

color

white to off-white

λ

conforms (UV/VIS Identity)

suitability

conforms to structure for H-NMR
conforms to structure for LC-MS

compatibility

configured for (ÄKTA® and OligoPilot®)

nucleoside profile

base: guanosine
base protecting group: isobutyryl
2' protecting group: TBDMS
5' protecting group: DMT
deprotection: standard

storage temp.

−20°C

SMILES string

COc1ccc(cc1)C(OC[C@H]2O[C@H]([C@H](O[Si](C)(C)C(C)(C)C)[C@@H]2OP(OCCC#N)N(C(C)C)C(C)C)n3cnc4C(=O)NC(NC(=O)C(C)C)=Nc34)(c5ccccc5)c6ccc(OC)cc6

InChI

1S/C50H68N7O9PSi/c1-32(2)45(58)54-48-53-44-41(46(59)55-48)52-31-56(44)47-43(66-68(12,13)49(7,8)9)42(65-67(63-29-17-28-51)57(33(3)4)34(5)6)40(64-47)30-62-50(35-18-15-14-16-19-35,36-20-24-38(60-10)25-21-36)37-22-26-39(61-11)27-23-37/h14-16,18-27,31-34,40,42-43,47H,17,29-30H2,1-13H3,(H2,53,54,55,58,59)/t40-,42-,43-,47-,67?/m1/s1

InChI key

PGJKESPHANLWME-FTZVBZPOSA-N

Looking for similar products? Visit Product Comparison Guide

General description

DMT-2′O-TBDMS-rG(ib) Phosphoramidite belongs to the group of 2′O-TBDMS RNA Phosphoramidites.

Application

RNA interference (RNAi) has become a popular tool for the sequence-specific inhibition of gene expression and can be used in target validation and other drug development techniques. The most convenient method to provide sequence-specific RNA oligonucleotides is a chemical synthesis on a solid support with RNA phosphoramidites and RNA CPG, analogous to DNA synthesis.

Features and Benefits

  • Industry-standard 2′O-TBDMS protective group
  • Consistent lot-to-lot purity and performance
  • Compatible with deprotection methods based on methylamine or AMA
  • Standard RNA phosphoramidites provide excellent coupling results when used with ETT or BTT as an activator; best results are obtained with Activator 42
  • Capping with standard acetic anhydride capping reagent rather than with Fast Deprotection Cap A
  • Manufactured under a certified ISO 9001 quality system

Other Notes

The unique properties of RNA have stimulated the development of a variety of applications in diagnostics and therapeutics, as well as in basic molecular biology research where RNA can be used as:
  • Catalytic agents (ribozymes)
  • Affinity ligands (aptamers)
  • Agents to induce gene silencing (RNA interference) (a)

Legal Information

OligoPilot is a registered trademark of Cytiva
ÄKTA is a registered trademark of Cytiva

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service