Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

G115000

Sigma-Aldrich

DMT-dG(dmf) Phosphoramidite

≥99.0% (reversed phase HPLC)

Synonym(s):

DMT-dG(dmf) Amidite, N-[(dimethylamino)methylene]-5′-O-[bis(4-methoxyphenyl)phenylmethyl]-2′-deoxyguanosine, 3′-[2-cyanoethyl N,N-bis(1-methylethyl)phosphoramidite], N2-Dimethylformamidine-5′-O-(4,4′-dimethoxytrityl)-2′-deoxyguanosine-3′-O-[O-(2-cyanoethyl)-N,N′-diisopropylphosphoramidite]

Sign Into View Organizational & Contract Pricing

Select a Size

6 X 5 G
$767.00
6 X 10 G
$1,410.00
100 G
$2,040.00
6 X 20 G
$2,630.00
500 G
$17,050.00
1 KG
$26,430.00

$767.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
6 X 5 G
$767.00
6 X 10 G
$1,410.00
100 G
$2,040.00
6 X 20 G
$2,630.00
500 G
$17,050.00
1 KG
$26,430.00

About This Item

Empirical Formula (Hill Notation):
C43H53N8O7P
CAS Number:
Molecular Weight:
824.90
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.51

$767.00


Please contact Customer Service for Availability

Request a Bulk Order

type

for DNA synthesis

Quality Level

product line

Proligo Reagents

Assay

≥99% (31P-NMR)
≥99.0% (reversed phase HPLC)

form

powder

mol wt

824.90 g/mol

technique(s)

oligo synthesis: suitable

color

white to off-white

λ

conforms (UV/VIS Identity)

nucleoside profile

base: deoxyguanosine
base protecting group: DMF
2' protecting group: none
5' protecting group: DMT
deprotection: fast

storage temp.

-10 to -25°C

SMILES string

COc1ccc(cc1)C(OC[C@H]2O[C@H](C[C@@H]2OP(OCCC#N)N(C(C)C)C(C)C)n3cnc4C(=O)NC(\N=C/N(C)C)=Nc34)(c5ccccc5)c6ccc(OC)cc6

InChI

1S/C43H53N8O7P/c1-29(2)51(30(3)4)59(56-24-12-23-44)58-36-25-38(50-28-45-39-40(50)47-42(48-41(39)52)46-27-49(5)6)57-37(36)26-55-43(31-13-10-9-11-14-31,32-15-19-34(53-7)20-16-32)33-17-21-35(54-8)22-18-33/h9-11,13-22,27-30,36-38H,12,24-26H2,1-8H3,(H,47,48,52)/b46-27-/t36-,37+,38+,59?/m0/s1

InChI key

YRQAXTCBMPFGAN-UJASEYITSA-N

Looking for similar products? Visit Product Comparison Guide

General description

DMT-dG(dmf) Phosphoramidite belongs to fast deprotection phosphoramidites where the deprotection step of automated oligonucleotide synthesis is integral to synthesis time and final product quality. Changing the dG protecting group to the dimethylformamidine (dmf) baseprotecting group enables the rapid synthesis of high-purity, high-yield oligonucleotide, thus increasing the efficiency of high-throughput production.

Features and Benefits

Key Features of dG(dmf):
  • dG(dmf) is deprotected faster than the conventional dG(ib): the deprotection time in concentrated ammonia is reduced to 2 hours at 55 °C or 1 hour at 65 °C
  • The dG(dmf)-monomer is especially suitable for G-rich sequences: incomplete deprotection is greatly reduced in comparison with the conventional dG(ib)-monomer
  • dG(dmf)-phosphoramidite is as stable in solution as the standard dA(bz)-, dC(bz)- and dT-phosphoramidites
  • dG(dmf)-phosphoramidite can directly substitute for dG(ib)-phosphoramidite
  • No change is required in the reagents commonly used for DNA synthesis (Except a low concentration iodine oxidizer i.e., 0.02 M in iodine, should be employed)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service