Skip to Content
MilliporeSigma
All Photos(1)

Documents

F6812

Sigma-Aldrich

Fluoroshield with PPD

liquid

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12171500
NACRES:
NA.47

product name

Fluoroshield with PPD,

form

liquid

Quality Level

pH range

8-8.4

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

2-8°C

Application

Fluoroshield with PPD (p-phenylenediamine; 1,4-phenylenediamine) is an aqueous mounting medium for preserving fluorescence of tissue and cell smears. Fluoroshield prevents rapid photobleaching of FITC, Texas Red®, AMCA, Alexa Fluor 488, Alexa Fluor 594, and tetramethylrhodamine. This mounting medium is fortified with PPD, an anti-fade reagent used to retard photobleaching of fluorescein and other fluorescent dyes. NOTE: Fluoroshield with PPD is NOT SUITABLE for immunofluorescence techniques involving Cy® dyes, R-phycoerythrin (R-PE), phycocyanin (PC),or allophycocyanin (APC).

Legal Information

ALEXA FLUOR is a trademark of Life Technologies
Cy is a registered trademark of Cytiva
FluoroShield is a trademark of ImmunoBioScience Corp.
Texas Red is a registered trademark of Life Technologies

related product

Product No.
Description
Pricing

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service