Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

S7112

Sigma-Aldrich

Propidium Iodide/Antifade Solution

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
$273.00

$273.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
$273.00

About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.41

$273.00


Please contact Customer Service for Availability

Request a Bulk Order

manufacturer/tradename

Chemicon®

Quality Level

technique(s)

activity assay: suitable (apoptosis)

detection method

fluorometric

shipped in

dry ice

General description

Propidium Iodide: {3,8-diamino-5-[3-(diethylmethylammonio)propyl]-6-phenylphenanthridinium diodide}. Antifade Solution: p-Phenylenediamine and glycerol in PBS solution. 2.5μg/mL Propidium Iodide (PI) suspended in Antifade.

Application

Propidium Iodide is a nuclear fluorescent stain used for counterstaining paraffin-embedded, formalin-fixed tissue, to discriminate between live vs. dead cells in flow cytometry, and staining metaphase chromosomes or interphase nuclei that have been hybridized with probes detected with Fluorescein, or equivalent fluors. Antifade prevents the permanent loss of fluorescence or fluorescent signal quenching due to photobleaching and prolonged exposure to high intensity light sources.



Materials Provided

Each vial contains 1 ml of Propidium Iodide/Antifade Solution.



Warnings and Precautions

Propidium Iodide is a potential carcinogen. Antifade is a potential toxin and irritant. Use extreme caution. Avoid contact with eyes, skin and mucous membranes. Wear gloves and a lab coat when handling. Follow your institution′s guidelines for hazardous waste disposal. Keep tightly capped and limit exposure to light.



Related Products

Antifade Solution S7114

DAPI/Antifade Solution S7113

Propidium Iodide Solution S7109

Storage and Stability

Recommended Storage

-15ºC to -25ºC

Protect from light

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service