Key Documents
EPS41I1200100
Eppendorf® New Brunswick S41i CO2 Incubator Shaker
CO2 incubator shaker with inner shelf and touch screen control, 170L, 1 (2nd optional) shelves, AC/DC input 110 V, 50 - 60 Hz, diam. 2.5 cm , Orbit Diameter 1in
Synonym(s):
Eppendorf® New Brunswick S41i
About This Item
UNSPSC Code:
41106300
NACRES:
NB.73
feature
1 (2nd optional) shelves
CO2 incubator shaker with inner shelf and touch screen control
manufacturer/tradename
Eppendorf® S41I-120-0100
diam.
2.5 cm , Orbit Diameter 1in
capacity
170L
General description
The New Brunswick S41i is the only CO2 incubator with an Eppendorf shaker inside. The S41i precisely controls temperature, shaking speed, and CO2, providing the ideal environment for growing mammalian and stem cells in culture. Accepts flasks up to 4 liters. Standard perforated shelf allows for static incubation at the same time as shaking and a second static shelf can be added as an option. Renowned triple eccentric drive provides stable, uniform and vibration-free motion connected to the shaking platform by four encapsulated support rods. The drive mechanism located below the chamber ensures that heat generated by the drive does not affect the critical environment within the chamber. The intuitive touch screen display and controls allows you to operate and monitor performance.
Features and Benefits
  • Temperature range from 4°c above ambient to 50°c
  • Shaking speeds between 25-400 rpm, 25-300 rpm if stacked, provides versatility for culturing a wide range of cell types
  • Large 35.6 x 61 cm (14 in x 24 in) platform accommodates flasks up to 4 liters providing flexibility with your cultures
  • Single or double stacked for increased capacity
  • Inner and outer door saves valuable gasses while maintaining the perfect enviornment for your cells
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
New Brunswick is a trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)