Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EP5895170005

Rotor for Eppendorf® 5910 R Centrifuge

S-4x500, incl. 4 x 500 mL rectangular buckets

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$5,080.00

$5,080.00


Please contact Customer Service for Availability

Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
1 EA
$5,080.00

About This Item

UNSPSC Code:
41103909
NACRES:
NB.82

$5,080.00


Please contact Customer Service for Availability

Notify Me

Get notified when this item is ready to ship via email.

feature

to hold x plates
to hold 0.2 mL to 500 mL tues and bottles
S-4x500, incl. 4 x 500 mL rectangular buckets

packaging

pack of 1 ea

manufacturer/tradename

Eppendorf® 5895170005

parameter

4,000 max. RCF (3,220 xg)

technique(s)

centrifugation: suitable

compatibility

for use with centrifuge models 5910R

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Rotor for Eppendorf(R)5910R Centrifuge

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service