EP4430603006
Shelf Rack for Eppendorf® Easypet 3
to hold, 1 Easypet 3
Synonym(s):
Accessories for Eppendorf® Easypet® 3
Select a Size
$42.90
Select a Size
About This Item
$42.90
Recommended Products
description
shelf rack
feature
to hold 1 Easypet 3
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 4430603006
Looking for similar products? Visit Product Comparison Guide
Application
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service