Key Documents
EP0030124332
Eppendorf® PCR tubes
colorless, capacity 0.2 mL, pack of 1000 ea
Synonym(s):
eppendorf tubes, microcentrifuge tubes, microfuge tubes
About This Item
UNSPSC Code:
41121800
NACRES:
NC.21
Product EP0030124332 is not currently sold in your country. Contact Technical Service
material
cap (plug seal)
colorless
colorless cap
colorless tube
conical bottom
feature
graduations: no
neck thread Push fit
packaging
pack of 1000 ea
manufacturer/tradename
Eppendorf® 0030124332
capacity
0.2 mL
Features and Benefits
  • The hinged lid with a defined position and the tight sealing prevents contamination and evaporation.
  • The lid is etched to ensure labeling is convenient.
  • Designed to have maximum transparency even on the tube base
  • Easy to open
  • Suitable with all thermal cyclers with 0.2 mL block format
  • The product is batch-tested and certified free of DNA, DNase, RNase and PCR inhibitors (PCR clean).
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)