Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EP0030122178

Eppendorf® Conical Tubes

sterile, 50 mL, colorless, pkg of 500 tubes (10 bags of 50 tubes)

Synonym(s):

eppendorf tubes, microcentrifuge tubes, microfuge tubes

Sign Into View Organizational & Contract Pricing

Select a Size

500 EA
$418.00

$418.00


Please contact Customer Service for Availability


Select a Size

Change View
500 EA
$418.00

About This Item

UNSPSC Code:
41121703
NACRES:
NB.22

$418.00


Please contact Customer Service for Availability

material

conical bottom polypropylene

sterility

sterile

feature

DNA free
RNase and DNase free
endotoxin free
with screw cap

packaging

pkg of 500 tubes (10 bags of 50 tubes)

manufacturer/tradename

Eppendorf® 0030122178

measuring range

50 mL

parameter

19500 × g max. RCF

D

30 mm

color

colorless

Looking for similar products? Visit Product Comparison Guide

General description

  • Eppendorf® conical tubes are manufactured from high-quality chemically resistant polypropylene (PP).
  • Ideal for most cell culture and cell biology applications.
  • Ergonomic design - one-handed operation for easy opening/closing.
  • Free of detectable slip agents, plasticizers, and biocides ensure optimal sample integrity.
  • Pyrogen-, DNase-, RNase-, human and bacterial DNA-free for convenient and safe cell culture applications.
  • Cap with flattened and grooved sides minimizes contamination and evaporation risks.
  • A large writing area on the tube facilitates accurate labeling.
  • Compatible with centrifuge rotors, mixers, and shakers.

Application

  • Sample preparation in microbiology and molecular biology labs
  • Large volume storage of samples and solutions
  • Preparation of master mixes and buffers

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service