Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EP0030121155

Eppendorf® Safe-Lock micro test tubes

capacity 0.5 mL, amber, pack of 500 ea

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121702
NACRES:
NB.11
Product EP0030121155 is not currently sold in your country. Contact Technical Service

material

amber
amber cap
amber tube
cap (plug seal)
conical bottom
polypropylene cap
polypropylene tube

sterility

non-sterile

feature

graduations: no
neck thread Push fit

packaging

pack of 500 ea

manufacturer/tradename

Eppendorf® 0030121155

capacity

0.5 mL

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

  • The hinged Eppendorf® Safe-Lock lid provides additional protection against unintentional lid opening during incubation and storage.
  • g-Safe®:Provides great centrifugation stability and prevents sample loss due to tube breakage
  • The optimized sealing properties ensures safety when working with hazardous samples and minimal evaporation rates during long-term storage.
  • The large frosted lid and wide side-surface area make labeling convenient.
  • Functions between temperatures -86 °C to 100 °C
  • Autoclavable when open (121 °C, 20 min)
Eppendorf® Safe-Lock tubes protect the sample with excellent centrifugation stability, feature optimized sealing properties, and reliably prevent sample evaporation. The features include:
The features include:
  • Hinged Eppendorf Safe-Lock lid prevents unintentional lid opening during incubation and storage.
  • Polypropylene provides high resistance to chemicals, mechanical stress and temperature extremes.
  • Manufactured without the use of slip agents, plasticizers, and biocides – substances that have been shown to leach from plastic consumables into the sample and negatively affect bio-assay results, g-Safe®: Exceptional centrifugation stability up to 30,000 × g prevents sample loss due to tube breakage and provides extra safety when working with hazardous samples.
  • Precise lid sealing for minimal evaporation rates during long-term storage.
  • Large frosted lid and surface on the side for easy labelling.
  • Guaranteed functionality from -86 °C to 100 °C.
  • Autoclavable when open (121 °C, 20 min).
  • Available in Eppendorf Protein and DNA LoBind material for maximum recovery of valuable samples.


Using the Eppendorf 30,000 × g system comprising of Safe-Lock Tubes and Centrifuge 5430 R with high-speed rotor, the effect of relative centrifugal force (rcf), duration of centrifugation and the amount of isopropanol on the recovery rate of plasmid DNA from alcohol precipitation were investigated. All three factors play a role, with rcf having the greatest effect. By centrifuging the Eppendorf Safe-Lock Tubes at 30,000 × g, nearly 90% of DNA could be recovered from a 5 min centrifugation.

Legal Information

Eppendorf is a registered trademark of Eppendorf AG
g-Safe is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service