Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EHU149811

Sigma-Aldrich

MISSION® esiRNA

targeting human IGFBP5

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$303.00
50 μG
$571.00

$303.00


Please contact Customer Service for Availability


Select a Size

Change View
20 μG
$303.00
50 μG
$571.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

$303.00


Please contact Customer Service for Availability

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

AGTGTGGGCTTTTTCCCTTTTTTGCTCCTTTTCATTACCCCTCCTCCGTTTTCACCCTTCTCCGGACTTCGCGTAGAACCTGCGAATTTCGAAGAGGAGGTGGCAAAGTGGGAGAAAAGAGGTGTTAGGGTTTGGGGTTTTTTTGTTTTTGTTTTTGTTTTTTAATTTCTTGATTTCAACATTTTCTCCCACCCTCTCGGCTGCAGCCAACGCCTCTTACCTGTTCTGCGGCGCCGCGCACCGCTGGCAGCTGAGGGTTAGAAAGCGGGGTGTATTTTAGATTTTAAGCAAAAATTTTAAAGATAAATCCATTTTTCTCTCCCACCCCCAACGCCATCTCCACTGCATCCGATCTCATTATTTCGGTGGTTGCTTGGGGGTGAACAATTTTGTGGCTTTTTTTCCCCTATAATTCTGACCCGCTCAGGCTTGAGGGTTTCTCCGGCCTCCGCTCACTGCGTGCACCTGGCGCTGCCCTGCTTCCCCCAACCTGTTGCAAGGCTTTAATTCTTGCAACTGGGACCTG

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dong Hyup Lee et al.
The Korean journal of physiology & pharmacology : official journal of the Korean Physiological Society and the Korean Society of Pharmacology, 17(2), 157-162 (2013-04-30)
Insulin-like growth factor binding proteins (IGFBPs) are important components of insulin growth factor (IGF) signaling pathways. One of the binding proteins, IGFBP-5, enhances the actions of IGF-1, which include the enhanced proliferation of smooth muscle cells. In the present study
Younghay Lee et al.
Cells, 8(4) (2019-04-26)
Type 2 diabetes mellitus (T2DM) is a prevalent chronic metabolic disorder accompanied by high blood glucose, insulin resistance, and relative insulin deficiency. Endoplasmic reticulum (ER) stress induced by high glucose and free fatty acids has been suggested as one of
Bong-Ki Hong et al.
Experimental & molecular medicine, 49(8), e363-e363 (2017-08-05)
Fibroblast-like synoviocytes (FLSs) constitute a major cell subset of rheumatoid arthritis (RA) synovia. Dysregulation of microRNAs (miRNAs) has been implicated in activation and proliferation of RA-FLSs. However, the functional association of various miRNAs with their targets that are characteristic of
Wenyu Wang et al.
Molecular cancer therapeutics, 17(9), 1973-1983 (2018-06-22)
Despite showing promise against PIK3CA-mutant breast cancers in preclinical studies, PI3K/AKT pathway inhibitors demonstrate limited clinical efficacy as monotherapy. Here, we found that histone H3K27me3 demethylase KDM6B-targeted IGFBP5 expression provides a protective mechanism for PI3K/AKT inhibitor-induced apoptosis in breast cancer
Junyun Wang et al.
Oncotarget, 6(24), 20636-20649 (2015-05-27)
The insulin-like growth factor binding protein 5 (IGFBP5), which is often dysregulated in human cancers, plays a crucial role in carcinogenesis and cancer development. However, the function and underlying mechanism of IGFBP5 in tumor growth and metastasis has been elusive

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service