MilliporeSigma
  • MicroRNA-143 and -145 modulate the phenotype of synovial fibroblasts in rheumatoid arthritis.
MicroRNA-143 and -145 modulate the phenotype of synovial fibroblasts in rheumatoid arthritis.
Experimental & molecular medicine (2017-08-05)
Bong-Ki Hong, Sungyong You, Seung-Ah Yoo, Dohyun Park, Daehee Hwang, Chul-Soo Cho, Wan-Uk Kim
ABSTRACT
Fibroblast-like synoviocytes (FLSs) constitute a major cell subset of rheumatoid arthritis (RA) synovia. Dysregulation of microRNAs (miRNAs) has been implicated in activation and proliferation of RA-FLSs. However, the functional association of various miRNAs with their targets that are characteristic of the RA-FLS phenotype has not been globally elucidated. In this study, we performed microarray analyses of miRNAs and mRNAs in RA-FLSs and osteoarthritis FLSs (OA-FLSs), simultaneously, to validate how dysregulated miRNAs may be associated with the RA-FLS phenotype. Global miRNA profiling revealed that miR-143 and miR-145 were differentially upregulated in RA-FLSs compared to OA-FLSs. miR-143 and miR-145 were highly expressed in independent RA-FLSs. The miRNA-target prediction and network model of the predicted targets identified insulin-like growth factor binding protein 5 (IGFBP5) and semaphorin 3A (SEMA3A) as potential target genes downregulated by miR-143 and miR-145, respectively. IGFBP5 level was inversely correlated with miR-143 expression, and its deficiency rendered RA-FLSs more sensitive to TNFα stimulation, promoting IL-6 production and NF-κB activity. Moreover, SEMA3A was a direct target of miR-145, as determined by a luciferase reporter assay, antagonizing VEGF
Sigma-Aldrich
MISSION® esiRNA, targeting human IGFBP5
Sigma-Aldrich
DL-Cysteine, technical grade