Skip to Content
MilliporeSigma
All Photos(1)

Documents

EHU041431

Sigma-Aldrich

MISSION® esiRNA

targeting human AQP4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GCATGTGATTGACGTTGACCGGGGAGAGGAGAAGAAGGGGAAAGACCAATCTGGAGAGGTATTGTCTTCAGTATGACTAGAAGATCGCACTGAAAGCAGACAAGACTCCTTAGAACTGTCCTCAGATTTCCTTCCACCCATTAAGGAAACAGATTTGTTATAAATTAGAAATGTGCAGGTTTGTTGTTTCATGTCATATTACTCAGTCTAAACAATAAATATTTCATAATTTACAAAGGAGGAACGGAAGAAACCTATTGTGAATTCCAAATCTAAAAAAAGAAATATTTTTAAAATGTTCTTAAGCAAATATATACCTATTTTATCTAGTTACCTTTCATTAACAACCAATTTTAACCGTGTGTCAAGATTTGGTTAAGTCTTGCCTGACAGAACTCAAAGACACG

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Letizia Granieri et al.
PloS one, 7(6), e38896-e38896 (2012-06-22)
Neuromyelitis optica (NMO) is a severely disabling autoimmune disorder of the central nervous system, which predominantly affects the optic nerves and spinal cord. In a majority of cases, NMO is associated with antibodies to aquaporin-4 (AQP4) (termed NMO-IgG). In this
Chao Yang et al.
IUBMB life, 67(3), 182-190 (2015-04-11)
Emerging evidence indicates that the water channel protein aquaporin 4 (AQP4) plays an essential role in water homeostasis and is implicated in the pathogenesis of brain edema. This study aimed to understand the physiological role of AQP4 in hypoxia-ischemia-mediated cytotoxic
Yusi Cheng et al.
Clinical and experimental pharmacology & physiology, 44(11), 1106-1115 (2017-07-09)
Aquaporin 4 (AQP4) is a type of water channel protein that maintains the water balance of cardiomyocytes. However, the physiological role of AQP4 in cardiovascular disease is poorly understood. We wanted to explore whether p66Shc and endoplasmic reticulum stress participates
Jing Jin et al.
European neurology, 83(6), 581-590 (2020-11-02)
Stroke is one of the leading causes of mortality and disability worldwide. Long noncoding RNAs (lncRNAs) including MALAT1 have been shown to have critical roles in cerebral ischemia reperfusion injury (CIRI). However, the underlying mechanism of MALAT1 in CIRI has
Dongfeng Niu et al.
PloS one, 7(7), e40770-e40770 (2012-07-19)
Aquaporin3 (AQP3) and Aquaporin4 (AQP4) play a major role in transcellular and transepithelial water movement as water channel membrane proteins. Little is known of their expression and significance in human thyroid tissues. Thus, we examined the expression of AQP3 and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service